Assay Details
Target Gene Details
Entrez Gene ID: | 6002 |
Gene Name: | regulator of G-protein signaling 12 |
Gene Aliases: |
- |
Location: |
Chr.4:3292978-3439913 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS12 | NM_002926.3 | 2 | 2393 | NP_002917.1 |
| NM_198229.2 | 2 | 2393 | NP_937872.1 | |
| XM_006713905.1 | 2 | 1712 | XP_006713968.1 | |
| XM_006713906.2 | 2 | 1712 | XP_006713969.1 | |
| XM_011513543.2 | XP_011511845.1 | |||
| XM_017008529.1 | 2 | 2962 | XP_016864018.1 | |
| XM_017008530.1 | 2 | 1712 | XP_016864019.1 | |
| XM_017008531.1 | 1 | 909 | XP_016864020.1 | |
| AF030111.1 | 2 | 2392 | AAB84114.1 | |
| AF035152.1 | 1 | 1543 | AAC39835.1 | |
| AF464735.1 | 1 | 1592 | AAL69959.1 | |
| AF464736.1 | 1 | 1592 | AAL69960.1 | |
| AK293767.1 | 2 | 1645 | ||
| AK310999.1 | ||||
| BC028154.1 | 2 | 1655 | AAH28154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461165 | Chr.4:3281695 - 3340410 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map