Assay Details
Target Gene Details
Entrez Gene ID: | 201626 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, 2-PDE |
Location: |
Chr.3:57556247-57656480 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE12 | NM_001322176.1 | 1 | 1204 | NP_001309105.1 |
| NM_001322177.1 | 1 | 1204 | NP_001309106.1 | |
| NM_177966.6 | 1 | 1204 | NP_808881.3 | |
| AB115695.1 | 1 | 1142 | BAD20938.1 | |
| AK074423.1 | 1 | 1153 | BAB85079.1 | |
| AK300374.1 | 1 | 1177 | ||
| AL831824.1 | 1 | 708 | CAD38538.1 | |
| BM713922.1 | 1 | 227 | ||
| BX647169.1 | 1 | 1174 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590368 | Chr.3:57483370 - 57576753 on Build GRCh38 | Gain |
|
| nsv1008684 | Chr.3:57537986 - 57766490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map