Assay Details
Target Gene Details
Entrez Gene ID: | 64326 |
Gene Name: | ring finger and WD repeat domain 2 |
Gene Aliases: |
COP1, RNF200 |
Location: |
Chr.1:175944826-176207279 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFWD2 | NM_001001740.3 | NP_001001740.1 | ||
| NM_001286644.1 | NP_001273573.1 | |||
| NM_022457.6 | NP_071902.2 | |||
| XM_005245447.2 | XP_005245504.1 | |||
| XM_005245448.2 | XP_005245505.1 | |||
| XM_006711487.2 | XP_006711550.1 | |||
| XM_011509881.1 | XP_011508183.1 | |||
| XM_017002076.1 | XP_016857565.1 | |||
| AF508940.1 | AAM34692.1 | |||
| AF527539.1 | AAQ08989.1 | |||
| AK001278.1 | ||||
| AK314750.1 | ||||
| AY509921.1 | AAS82851.1 | |||
| BC094728.1 | AAH94728.1 | |||
| BX537932.1 | ||||
| CA450176.1 | ||||
| KJ534928.1 | ||||
| KJ534929.1 | ||||
| KJ534930.1 | ||||
| KJ534931.1 | ||||
| KJ534932.1 | ||||
| KJ534933.1 | ||||
| KJ534992.1 | ||||
| KJ535061.1 | ||||
| KJ535076.1 |
More Information
Set Membership: |
|

Genomic Map