Assay Details
Target Gene Details
Entrez Gene ID: | 3693 |
Gene Name: | integrin subunit beta 5 |
Gene Aliases: |
- |
Location: |
Chr.3:124761948-124901411 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB5 | NM_002213.4 | NP_002204.2 | ||
| XM_005247436.2 | XP_005247493.1 | |||
| XM_006713630.2 | XP_006713693.1 | |||
| XM_017006351.1 | XP_016861840.1 | |||
| XM_017006352.1 | XP_016861841.1 | |||
| XM_017006353.1 | XP_016861842.1 | |||
| XM_017006354.1 | XP_016861843.1 | |||
| XM_017006355.1 | XP_016861844.1 | |||
| AK054968.1 | ||||
| AK130168.1 | ||||
| AK309551.1 | ||||
| AK315429.1 | ||||
| BC006541.2 | AAH06541.1 | |||
| J05633.1 | AAA59183.1 | |||
| M35011.1 | AAA52707.1 | |||
| X53002.1 | CAA37188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3332098 | Chr.3:124763415 - 124765313 on Build GRCh38 | Duplication |
|
| nsv1012242 | Chr.3:124722848 - 124797029 on Build GRCh38 | Gain |
|
| nsv1001677 | Chr.3:124015843 - 125108694 on Build GRCh38 | Loss |
|
| nsv1160974 | Chr.3:124577493 - 125300995 on Build GRCh38 | Duplication |
|
| nsv3981 | Chr.3:124757649 - 124788489 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map