Assay Details
Target Gene Details
Entrez Gene ID: | 5147 |
Gene Name: | phosphodiesterase 6D |
Gene Aliases: |
JBTS22, PDED |
Location: |
Chr.2:231732425-231781327 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6D | NM_001291018.1 | NP_001277947.1 | ||
| NM_002601.3 | NP_002592.1 | |||
| XM_011511342.1 | XP_011509644.1 | |||
| AF022912.1 | AAB87872.1 | |||
| BC007831.2 | AAH07831.1 | |||
| BI754373.1 | ||||
| BT007278.1 | AAP35942.1 | |||
| CR456980.1 | CAG33261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428408 | Chr.2:231672114 - 231968956 on Build GRCh38 | Gain+Loss |
|
| nsv834568 | Chr.2:231625367 - 231780141 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map