Assay Details
Target Gene Details
Entrez Gene ID: | 57214 |
Gene Name: | cell migration inducing hyaluronan binding protein |
Gene Aliases: |
CCSP1, HYBID, KIAA1199, TMEM2L |
Location: |
Chr.15:80779343-80951662 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEMIP | NM_001293298.1 | 1 | 152 | NP_001280227.1 |
| NM_001293304.1 | 1 | 152 | NP_001280233.1 | |
| NM_018689.2 | 1 | 152 | NP_061159.1 | |
| AB103330.1 | 1 | 124 | BAD02451.1 | |
| AK000465.1 | 1 | 124 | ||
| AY581148.1 | 1 | 124 | AAU06219.1 | |
| AY581149.1 | 1 | 124 | AAU06220.1 | |
| AY585237.1 | 1 | 124 | AAU06221.1 | |
| BC020256.1 | 1 | 100 | AAH20256.1 | |
| DA220969.1 | 1 | 157 |
More Information
Set Membership: |
|

Genomic Map