Assay Details
Target Gene Details
Entrez Gene ID: | 84245 |
Gene Name: | methylthioribose-1-phosphate isomerase 1 |
Gene Aliases: |
MRDI, MTNA, Ypr118w |
Location: |
Chr.19:13764492-13774282 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRI1 | NM_001031727.2 | NP_001026897.1 | ||
| NM_032285.2 | NP_115661.1 | |||
| XM_011528355.2 | XP_011526657.1 | |||
| XM_011528356.2 | XP_011526658.1 | |||
| XM_011528357.2 | XP_011526659.1 | |||
| AL834276.1 | CAD38951.1 | |||
| AY358176.1 | AAQ88543.1 | |||
| BC001703.1 | AAH01703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893166 | Chr.19:13629109 - 13774670 on Build GRCh38 | Gain |
|
| nsv833757 | Chr.19:13725904 - 13951988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map