Assay Details
Target Gene Details
Entrez Gene ID: | 9125 |
Gene Name: | CCR4-NOT transcription complex subunit 9 |
Gene Aliases: |
CAF40, CT129, RCD-1, RCD1, RQCD1 |
Location: |
Chr.2:218568579-218596435 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT9 | NM_001271634.1 | 9 | 1290 | NP_001258563.1 |
| NM_005444.2 | 8 | 1194 | NP_005435.1 | |
| NR_073390.1 | 7 | 1084 | ||
| XM_011512138.2 | 9 | 1313 | XP_011510440.1 | |
| XM_017005248.1 | 8 | 1783 | XP_016860737.1 | |
| AB500892.1 | 8 | 1066 | ||
| AK293281.1 | 9 | 1290 | ||
| AK296037.1 | 4 | 347 | ||
| AK316567.1 | 8 | 900 | ||
| BC137455.1 | 8 | 916 | ||
| BC137456.1 | 8 | 916 |
More Information
Set Membership: |
|

Genomic Map