Assay Details
Target Gene Details
Entrez Gene ID: | 51312 |
Gene Name: | solute carrier family 25 member 37 |
Gene Aliases: |
HT015, MFRN, MFRN1, MSC, MSCP, PRO1278, PRO1584, PRO2217 |
Location: |
Chr.8:23528795-23575463 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A37 | NM_001317812.1 | 1 | 316 | NP_001304741.1 |
| NM_001317813.1 | 1 | 316 | NP_001304742.1 | |
| NM_001317814.1 | 1 | 316 | NP_001304743.1 | |
| NM_016612.3 | 1 | 316 | NP_057696.2 | |
| AF223466.1 | 1 | 152 | AAF64141.1 | |
| AK074708.1 | 1 | 155 | ||
| AK075313.1 | 1 | 81 | ||
| AK223194.1 | 1 | 157 | BAD96914.1 | |
| AL833186.1 | CAH10415.1 | |||
| AY032628.1 | 1 | 152 | AAK38154.1 | |
| BC015013.2 | 1 | 121 | AAH15013.1 | |
| BC132799.1 | 1 | 180 | ||
| BC132801.1 | 1 | 180 | ||
| BI460337.1 | 1 | 270 | ||
| BM552840.1 | 1 | 251 | ||
| BM926580.1 | 1 | 159 | ||
| BU166030.1 | 1 | 236 | ||
| DB057758.1 | 1 | 316 | ||
| HY049859.1 | 1 | 259 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508499 | Chr.8:23452508 - 23565200 on Build GRCh38 | Deletion |
|
| nsv1017570 | Chr.8:22969562 - 23544783 on Build GRCh38 | Gain |
|
| nsv1026345 | Chr.8:23302920 - 23541056 on Build GRCh38 | Gain |
|
| nsv831268 | Chr.8:23357001 - 23538079 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map