Assay Details
Target Gene Details
Entrez Gene ID: | 8812 |
Gene Name: | cyclin K |
Gene Aliases: |
CPR4 |
Location: |
Chr.14:99481402-99511515 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNK | NM_001099402.1 | NP_001092872.1 | ||
| XM_005268154.4 | XP_005268211.1 | |||
| XM_011537275.2 | XP_011535577.1 | |||
| AK311075.1 | ||||
| BC015935.1 | AAH15935.1 | |||
| BT006950.1 | AAP35596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635481 | Chr.14:99277609 - 99600560 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map