Assay Details
Target Gene Details
Entrez Gene ID: | 57549 |
Gene Name: | immunoglobulin superfamily member 9 |
Gene Aliases: |
FP18798, IGSF9A, Nrt1 |
Location: |
Chr.1:159927039-159945661 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF9 | NM_001135050.1 | NP_001128522.1 | ||
| NM_020789.3 | NP_065840.2 | |||
| XM_017001884.1 | XP_016857373.1 | |||
| AB037776.1 | ||||
| AY358128.1 | AAQ88495.1 | |||
| BC030141.2 | AAH30141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548052 | Chr.1:159891127 - 159938603 on Build GRCh38 | Gain |
|
| nsv548053 | Chr.1:159915710 - 159938603 on Build GRCh38 | Loss |
|
| nsv516637 | Chr.1:159919896 - 159938603 on Build GRCh38 | Loss |
|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
| nsv548054 | Chr.1:159926189 - 159937651 on Build GRCh38 | Loss |
|
| nsv524006 | Chr.1:159916632 - 159953321 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map