Assay Details
Target Gene Details
Entrez Gene ID: | 8125 |
Gene Name: | acidic nuclear phosphoprotein 32 family member A |
Gene Aliases: |
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32 |
Location: |
Chr.15:68778535-68820922 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANP32A | NM_006305.3 | NP_006296.1 | ||
| AF025684.1 | AAB91548.1 | |||
| AK223280.1 | BAD97000.1 | |||
| AK312703.1 | ||||
| AW383928.1 | ||||
| AY349171.1 | AAQ79832.1 | |||
| BC000608.2 | AAH00608.1 | |||
| BC007200.1 | AAH07200.1 | |||
| BC106719.2 | AAI06720.1 | |||
| BC125143.1 | ||||
| BT007436.1 | AAP36104.1 | |||
| DA825897.1 | ||||
| U60823.1 | AAC50570.1 | |||
| U73477.1 | AAB39706.1 | |||
| X75090.1 | CAA52981.1 |
Target Gene Details
Entrez Gene ID: | 80035 |
Gene Name: | ANP32A intronic transcript 1 |
Gene Aliases: |
C15orf28, HsT18971, NCRNA00321 |
Location: |
Chr.15:68803821-68807101 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANP32A-IT1 | NR_026808.1 | 1 | 501 | |
| AK021784.1 | 1 | 501 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636811 | Chr.15:68805758 - 68813106 on Build GRCh38 | Loss |
|
| nsv833046 | Chr.15:68639374 - 68841420 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76895648] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map