Assay Details
Target Gene Details
Entrez Gene ID: | 2041 |
Gene Name: | EPH receptor A1 |
Gene Aliases: |
EPH, EPHT, EPHT1 |
Location: |
Chr.7:143390813-143408892 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA1 | NM_005232.4 | NP_005223.4 | ||
| XM_006715880.3 | XP_006715943.1 | |||
| AK290351.1 | ||||
| AK313263.1 | ||||
| BC130291.1 | ||||
| M18391.1 | AAA36747.1 | |||
| Z27409.1 | CAA81796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949967 | Chr.7:143382508 - 143399907 on Build GRCh38 | Deletion |
|
| nsv608681 | Chr.7:143372217 - 143425364 on Build GRCh38 | Loss |
|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map