Assay Details
Target Gene Details
Entrez Gene ID: | 79811 |
Gene Name: | SAFB like transcription modulator |
Gene Aliases: |
Met |
Location: |
Chr.15:58879045-58934313 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLTM | NM_001013843.2 | 21 | 3146 | NP_001013865.1 |
| NM_024755.3 | 21 | 3200 | NP_079031.2 | |
| NR_135042.1 | 19 | 2533 | ||
| NR_135043.1 | 19 | 2643 | ||
| XM_006720686.3 | 19 | 3591 | XP_006720749.3 | |
| XM_006720690.1 | 17 | 2297 | XP_006720753.1 | |
| XM_011522022.1 | 21 | 3858 | XP_011520324.1 | |
| XM_011522023.1 | 21 | 3804 | XP_011520325.1 | |
| XM_011522024.1 | 20 | 3708 | XP_011520326.1 | |
| XM_011522025.1 | 20 | 3660 | XP_011520327.1 | |
| XM_011522026.2 | 20 | 3639 | XP_011520328.2 | |
| XM_011522027.1 | 19 | 3612 | XP_011520329.1 | |
| XM_011522028.1 | 19 | 3558 | XP_011520330.1 | |
| XM_011522029.2 | 21 | 3039 | XP_011520331.1 | |
| XM_011522031.2 | 18 | 2694 | XP_011520333.1 | |
| XM_017022576.1 | 20 | 2996 | XP_016878065.1 | |
| XM_017022577.1 | 20 | 3585 | XP_016878066.1 | |
| XM_017022578.1 | 19 | 3537 | XP_016878067.1 | |
| XM_017022579.1 | 21 | 3203 | XP_016878068.1 | |
| XM_017022580.1 | 20 | 2657 | XP_016878069.1 | |
| AK023275.1 | 11 | 1654 | BAB14502.1 | |
| AK055195.1 | 19 | 2436 | ||
| AK291423.1 | 19 | 2566 | ||
| AL834297.1 | 7 | 1095 | CAH56362.1 | |
| BC014944.1 | 14 | 2003 | AAH14944.1 | |
| BC046119.1 | 10 | 1495 | ||
| BC140851.1 | 21 | 3167 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| nsv1035640 | Chr.15:58813161 - 58898731 on Build GRCh38 | Gain |
|
| esv34110 | Chr.15:58657792 - 59089435 on Build GRCh38 | Loss |
|
| nsv94675 | Chr.15:58872682 - 58882181 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map