Assay Details
Target Gene Details
Entrez Gene ID: | 64600 |
Gene Name: | phospholipase A2 group IIF |
Gene Aliases: |
- |
Location: |
Chr.1:20139326-20150386 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2F | NM_022819.3 | NP_073730.3 | ||
| XM_011541955.1 | XP_011540257.1 | |||
| XM_011541956.1 | XP_011540258.1 | |||
| AF306566.1 | AAG50242.1 | |||
| AK093645.1 | BAC04210.1 | |||
| BX647458.1 | 2 | 3503 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv1003802 | Chr.1:20057935 - 20154347 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map