Assay Details
Target Gene Details
Entrez Gene ID: | 116173 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 5 |
Gene Aliases: |
CKLFSF5 |
Location: |
Chr.14:23376439-23379772 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM5 | NM_001037288.1 | 1 | 93 | NP_001032365.1 |
| NM_001288744.1 | 1 | 93 | NP_001275673.1 | |
| NM_001288745.1 | 1 | 93 | NP_001275674.1 | |
| NM_001288746.1 | 1 | 93 | NP_001275675.1 | |
| NM_138460.2 | 1 | 93 | NP_612469.1 | |
| XM_017020953.1 | XP_016876442.1 | |||
| XM_017020954.1 | 1 | 86 | XP_016876443.1 | |
| AF479262.2 | 1 | 94 | AAN73040.1 |
More Information
Set Membership: |
|

Genomic Map