Assay Details
Target Gene Details
Entrez Gene ID: | 9119 |
Gene Name: | keratin 75 |
Gene Aliases: |
K6HF, KB18, PFB |
Location: |
Chr.12:52424070-52434326 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT75 | NM_004693.2 | 9 | 2027 | NP_004684.2 |
| AK298960.1 | ||||
| Y17282.1 | 9 | 2023 | CAA76730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832410 | Chr.12:52321596 - 52506980 on Build GRCh38 | Gain |
|
| dgv1504n100 | Chr.12:52423017 - 52454608 on Build GRCh38 | Gain |
|
| nsv558928 | Chr.12:52401803 - 52437285 on Build GRCh38 | Loss |
|
| nsv832411 | Chr.12:52347803 - 52494080 on Build GRCh38 | Loss |
|
| nsv832412 | Chr.12:52390206 - 52545452 on Build GRCh38 | Loss |
|
| nsv520797 | Chr.12:52421681 - 52434655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map