Assay Details
Target Gene Details
Entrez Gene ID: | 10426 |
Gene Name: | tubulin gamma complex associated protein 3 |
Gene Aliases: |
104p, GCP3, Grip104, SPBC98, Spc98, Spc98p |
Location: |
Chr.13:112485005-112606469 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP3 | NM_001286277.1 | NP_001273206.1 | ||
| NM_001286278.1 | NP_001273207.1 | |||
| NM_006322.5 | NP_006313.1 | |||
| XM_005268293.1 | XP_005268350.1 | |||
| XM_011537462.2 | XP_011535764.1 | |||
| XM_011537463.2 | XP_011535765.1 | |||
| XM_011537464.2 | XP_011535766.1 | |||
| XM_017020323.1 | XP_016875812.1 | |||
| AF042378.1 | AAC39727.1 | |||
| AJ003061.1 | CAA05832.1 | |||
| AJ003062.1 | CAA05833.1 | |||
| AK302537.1 | ||||
| AK314785.1 | ||||
| BC013781.1 | AAH13781.1 | |||
| BC046634.2 | AAH46634.1 | |||
| BT009772.1 | AAP88774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053676 | Chr.13:112332274 - 112553333 on Build GRCh38 | Gain |
|
| nsv1044644 | Chr.13:112437023 - 113055517 on Build GRCh38 | Gain |
|
| esv3633580 | Chr.13:112394808 - 112549597 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map