Assay Details
Target Gene Details
Entrez Gene ID: | 23414 |
Gene Name: | zinc finger protein, FOG family member 2 |
Gene Aliases: |
DIH3, FOG2, SRXY9, ZC2HC11B, ZNF89B, hFOG-2 |
Location: |
Chr.8:105318859-105804539 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFPM2 | NM_012082.3 | 8 | 1862 | NP_036214.2 |
| XM_011516947.2 | 8 | 5461 | XP_011515249.1 | |
| XM_011516948.2 | 7 | 1763 | XP_011515250.1 | |
| AB073894.1 | 1 | 732 | ||
| AF119334.1 | 8 | 1863 | AAD49558.1 | |
| AK296997.1 | 8 | 2011 | ||
| AK303058.1 | 6 | 1633 | ||
| AK316426.1 | 6 | 1624 | ||
| BC109222.1 | 8 | 1863 | AAI09223.1 | |
| BC109223.1 | 8 | 1863 | AAI09224.1 |
Target Gene Details
Entrez Gene ID: | 102723356 |
Gene Name: | ZFPM2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:105780410-106060503 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFPM2-AS1 | NR_125796.1 | |||
| NR_125797.1 | ||||
| BF244158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831418 | Chr.8:105636228 - 105821105 on Build GRCh38 | Gain |
|
| nsv6339 | Chr.8:105776390 - 105809823 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map