Assay Details
Target Gene Details
Entrez Gene ID: | 22858 |
Gene Name: | intestinal cell (MAK-like) kinase |
Gene Aliases: |
ECO, LCK2, MRK |
Location: |
Chr.6:53001299-53061864 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICK | NM_014920.3 | 11 | 1819 | NP_055735.1 |
| NM_016513.4 | 12 | 1930 | NP_057597.2 | |
| XM_011514419.1 | 11 | 1840 | XP_011512721.1 | |
| XM_011514420.2 | 12 | 1982 | XP_011512722.1 | |
| XM_011514421.1 | 12 | 1921 | XP_011512723.1 | |
| XM_017010485.1 | 12 | 2214 | XP_016865974.1 | |
| XM_017010486.1 | 13 | 2093 | XP_016865975.1 | |
| XM_017010487.1 | 11 | 1759 | XP_016865976.1 | |
| XM_017010488.1 | 12 | 2620 | XP_016865977.1 | |
| XM_017010489.1 | 13 | 2135 | XP_016865978.1 | |
| XM_017010490.1 | 12 | 2005 | XP_016865979.1 | |
| XM_017010491.1 | 12 | 1966 | XP_016865980.1 | |
| XM_017010492.1 | 11 | 2010 | XP_016865981.1 | |
| AB023153.1 | 11 | 1706 | ||
| AF152469.1 | 11 | 1787 | AAG43364.1 | |
| AF225919.1 | 11 | 1649 | AAF37278.1 | |
| AK074892.1 | 12 | 1900 | ||
| BC136420.1 | 10 | 1480 | ||
| BC136421.1 | 10 | 1480 | ||
| BC152464.1 | 11 | 1706 |
More Information
Set Membership: |
|

Genomic Map