Assay Details
Target Gene Details
Entrez Gene ID: | 9495 |
Gene Name: | A-kinase anchoring protein 5 |
Gene Aliases: |
AKAP75, AKAP79, H21 |
Location: |
Chr.14:64465499-64474503 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP5 | NM_004857.3 | 2 | 271 | NP_004848.3 |
| AK315336.1 | 2 | 271 | ||
| BC057229.1 | 2 | 185 | AAH57229.1 | |
| BC131516.1 | 1 | 503 | ||
| DA942424.1 | 2 | 271 | ||
| M90359.1 | 1 | 1190 | AAA58363.1 |
Target Gene Details
Entrez Gene ID: | 7597 |
Gene Name: | zinc finger and BTB domain containing 25 |
Gene Aliases: |
C14orf51, KUP, ZNF46 |
Location: |
Chr.14:64447172-64505213 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB25 | NM_001304508.1 | NP_001291437.1 | ||
| XM_006720250.3 | XP_006720313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952852 | Chr.14:64465983 - 64475282 on Build GRCh38 | Duplication |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map