Assay Details
Target Gene Details
Entrez Gene ID: | 64773 |
Gene Name: | PC-esterase domain containing 1A |
Gene Aliases: |
C20orf81, FAM113A, bA12M19.1 |
Location: |
Chr.20:2835314-2841243 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCED1A | NM_001271168.1 | NP_001258097.1 | ||
| NM_022760.4 | 1 | 333 | NP_073597.2 | |
| XM_005260804.2 | XP_005260861.1 | |||
| XM_005260805.2 | 1 | 266 | XP_005260862.1 | |
| XM_005260806.3 | 1 | 266 | XP_005260863.1 | |
| AK026029.1 | BAB15328.1 | |||
| AK056353.1 | 1 | 317 | BAB71160.1 | |
| AK293638.1 | 1 | 106 | ||
| BC014247.2 | AAH14247.1 | |||
| BC051816.1 | 1 | 121 | AAH51816.1 | |
| BE302577.1 | ||||
| DB276752.1 | ||||
| DC421675.1 | 1 | 333 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645020 | Chr.20:2816405 - 2926274 on Build GRCh38 | Gain |
|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv522009 | Chr.20:2745032 - 3030542 on Build GRCh38 | Gain |
|
| nsv833895 | Chr.20:2730164 - 2882236 on Build GRCh38 | Loss |
|
| nsv3261 | Chr.20:2820663 - 2851754 on Build GRCh38 | Deletion |
|
| nsv952978 | Chr.20:2835355 - 2842754 on Build GRCh38 | Deletion |
|
| esv2762056 | Chr.20:2740745 - 2944353 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map