Assay Details
Target Gene Details
Entrez Gene ID: | 6932 |
Gene Name: | transcription factor 7 (T-cell specific, HMG-box) |
Gene Aliases: |
TCF-1 |
Location: |
Chr.5:134114703-134148229 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF7 | NM_003202.3 | 1 | 297 | NP_003193.2 |
| XM_006714678.3 | 1 | 305 | XP_006714741.1 | |
| XM_006714679.3 | 1 | 305 | XP_006714742.1 | |
| XM_006714680.3 | 1 | 305 | XP_006714743.1 | |
| XM_006714682.2 | 1 | 305 | XP_006714745.1 | |
| XM_006714684.2 | 1 | 305 | XP_006714747.1 | |
| XM_011543604.2 | 1 | 305 | XP_011541906.1 | |
| XM_011543606.1 | 1 | 305 | XP_011541908.1 | |
| AL832816.1 | ||||
| BC048769.1 | 1 | 297 | AAH48769.1 | |
| BC072023.1 | 1 | 232 | AAH72023.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112885 | Chr.5:134114709 - 134115309 on Build GRCh38 | Deletion |
|
| nsv599732 | Chr.5:134099223 - 134230401 on Build GRCh38 | Loss |
|
| nsv5011 | Chr.5:134083161 - 134116977 on Build GRCh38 | Insertion |
|
| nsv1073496 | Chr.5:134113208 - 134115809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map