Assay Details
Target Gene Details
Entrez Gene ID: | 54414 |
Gene Name: | sialic acid acetylesterase |
Gene Aliases: |
AIS6, CSE-C, CSEC, LSE, YSG2 |
Location: |
Chr.11:124635789-124676303 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAE | NM_001199922.1 | NP_001186851.1 | ||
| NM_170601.4 | NP_733746.1 | |||
| XM_017017930.1 | XP_016873419.1 | |||
| AF300796.1 | AAG15386.1 | |||
| AF303378.1 | AAG14897.1 | |||
| AK056093.1 | ||||
| AK092224.1 | ||||
| AL137496.1 | CAB70771.1 | |||
| BC068450.1 | AAH68450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045156 | Chr.11:124565356 - 124705852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map