Assay Details
Target Gene Details
Entrez Gene ID: | 301 |
Gene Name: | annexin A1 |
Gene Aliases: |
ANX1, LPC1 |
Location: |
Chr.9:73151731-73170393 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANXA1 | NM_000700.2 | 5 | 556 | NP_000691.1 |
| XM_011518609.1 | 5 | 483 | XP_011516911.1 | |
| XM_017014657.1 | 5 | 502 | XP_016870146.1 | |
| AB451274.1 | 4 | 348 | ||
| AB451401.1 | 4 | 348 | ||
| AK074480.1 | 4 | 1230 | ||
| AK296808.1 | 5 | 422 | ||
| BC001275.1 | 5 | 400 | AAH01275.1 | |
| BC035993.1 | 5 | 422 | AAH35993.1 | |
| BT019896.1 | 4 | 348 | AAV38699.1 | |
| BT019916.1 | 4 | 348 | AAV38719.1 | |
| BT019917.1 | 4 | 348 | AAV38720.1 | |
| CR407684.1 | 4 | 348 | CAG28612.1 | |
| CX870105.1 | 5 | 556 | ||
| X05908.1 | 5 | 422 | CAA29338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620661 | Chr.9:73148689 - 73332974 on Build GRCh38 | Gain |
|
| nsv6561 | Chr.9:73140727 - 73175683 on Build GRCh38 | Insertion |
|
| nsv526225 | Chr.9:72987987 - 73372263 on Build GRCh38 | Gain |
|
| esv3620662 | Chr.9:73155774 - 73254536 on Build GRCh38 | Gain |
|
| nsv614612 | Chr.9:72979640 - 73296530 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map