Assay Details
Target Gene Details
Entrez Gene ID: | 2152 |
Gene Name: | coagulation factor III, tissue factor |
Gene Aliases: |
CD142, TF, TFA |
Location: |
Chr.1:94529176-94541857 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F3 | NM_001178096.1 | 5 | 1818 | NP_001171567.1 |
| NM_001993.4 | 6 | 1978 | NP_001984.1 | |
| AF497569.1 | 5 | 1545 | AAO61151.1 | |
| AF497570.1 | 5 | 1545 | AAO61152.1 | |
| AF497571.1 | 3 | 1318 | AAO61153.1 | |
| BM981095.1 | 1 | 293 | ||
| J02931.1 | 6 | 1868 | AAA61150.1 | |
| M16553.1 | 6 | 1832 | AAA61151.1 | |
| M27436.1 | 6 | 1856 | AAA36734.1 | |
| X07680.1 | 2 | 1115 | CAA30527.1 |
More Information
Set Membership: |
|

Genomic Map