Assay Details
Target Gene Details
Entrez Gene ID: | 10095 |
Gene Name: | actin related protein 2/3 complex subunit 1B |
Gene Aliases: |
ARC41, p40-ARC, p41-ARC |
Location: |
Chr.7:99374675-99394801 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC1B | NM_005720.3 | NP_005711.1 | ||
| XM_006715825.1 | XP_006715888.1 | |||
| XM_006715826.1 | XP_006715889.1 | |||
| AF006084.1 | AAB64189.1 | |||
| AK314818.1 | ||||
| AU143525.1 | ||||
| BC002562.2 | AAH02562.1 | |||
| BC002988.1 | AAH02988.2 | |||
| BC007555.1 | AAH07555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv831068 | Chr.7:99221949 - 99408858 on Build GRCh38 | Loss |
|
| nsv951379 | Chr.7:99375678 - 99405377 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map