Assay Details
Target Gene Details
Entrez Gene ID: | 255762 |
Gene Name: | PDZ domain containing 9 |
Gene Aliases: |
C16orf65 |
Location: |
Chr.16:21957540-22001137 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZD9 | NM_173806.3 | 3 | 629 | NP_776167.2 |
| NR_033694.1 | 5 | 880 | ||
| XM_006721029.3 | 4 | 833 | XP_006721092.1 | |
| XM_011545785.2 | 4 | 836 | XP_011544087.1 | |
| XM_017023109.1 | 3 | 655 | XP_016878598.1 | |
| XM_017023110.1 | 5 | 894 | XP_016878599.1 | |
| BC021724.1 | 5 | 885 | ||
| BC028605.1 | 5 | 885 | ||
| BC039562.1 | 5 | 877 | AAH39562.1 | |
| BX100380.1 | 1 | 150 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34235 | Chr.16:21276264 - 21986230 on Build GRCh38 | Gain |
|
| nsv1048054 | Chr.16:21952632 - 21994369 on Build GRCh38 | Gain |
|
| nsv428322 | Chr.16:21489241 - 22017793 on Build GRCh38 | Gain |
|
| esv3638193 | Chr.16:21934532 - 22029031 on Build GRCh38 | Gain |
|
| esv34544 | Chr.16:21522978 - 21986230 on Build GRCh38 | Gain |
|

Genomic Map