Assay Details
Target Gene Details
Entrez Gene ID: | 11057 |
Gene Name: | abhydrolase domain containing 2 |
Gene Aliases: |
HS1-2, LABH2, PHPS1-2 |
Location: |
Chr.15:89088150-89202360 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD2 | NM_007011.7 | 1 | 61 | NP_008942.3 |
| NM_152924.4 | 1 | 61 | NP_690888.1 | |
| DB024314.1 | 1 | 37 | ||
| DB049736.1 | ||||
| DC397201.1 | 1 | 61 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637182 | Chr.15:89038688 - 89124261 on Build GRCh38 | Loss |
|
| nsv1050740 | Chr.15:89006295 - 89155865 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map