Assay Details
Target Gene Details
Entrez Gene ID: | 4722 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S3 |
Gene Aliases: |
CI-30 |
Location: |
Chr.11:47579010-47584563 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS3 | NM_004551.2 | NP_004542.1 | ||
| AA477836.1 | ||||
| AF067139.1 | AAC27451.1 | |||
| AF100743.1 | AAD40386.1 | |||
| AK223255.1 | BAD96975.1 | |||
| AK313802.1 | ||||
| AL135819.1 | CAB63881.1 | |||
| BC000617.2 | AAH00617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
| nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map