Assay Details
Target Gene Details
Entrez Gene ID: | 5032 |
Gene Name: | purinergic receptor P2Y11 |
Gene Aliases: |
P2Y11 |
Location: |
Chr.19:10111521-10115389 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY11 | NM_002566.4 | 2 | 1618 | NP_002557.2 |
| BC073827.1 | 2 | 1601 | AAH73827.1 |
Target Gene Details
Entrez Gene ID: | 692312 |
Gene Name: | PPAN-P2RY11 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:10106223-10115389 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPAN-P2RY11 | NM_001040664.2 | 13 | 2849 | NP_001035754.1 |
| NM_001198690.1 | 13 | 2911 | NP_001185619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map