Assay Details
Target Gene Details
Entrez Gene ID: | 101929664 |
Gene Name: | HORMAD2 antisense RNA 1 |
Gene Aliases: |
MTMR3-AS1 |
Location: |
Chr.22:30008742-30080480 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HORMAD2-AS1 | NR_110541.1 | |||
| BG618381.1 |
Target Gene Details
Entrez Gene ID: | 8897 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
FYVE-DSP1, ZFYVE10 |
Location: |
Chr.22:29883165-30030868 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR3 | NM_021090.3 | NP_066576.1 | ||
| NM_153050.2 | NP_694690.1 | |||
| NM_153051.2 | NP_694691.1 | |||
| XM_005261803.2 | XP_005261860.1 | |||
| XM_005261804.3 | XP_005261861.1 | |||
| XM_005261805.2 | XP_005261862.1 | |||
| XM_005261806.2 | XP_005261863.1 | |||
| XM_005261808.2 | XP_005261865.1 | |||
| XM_017029028.1 | XP_016884517.1 | |||
| XM_017029029.1 | XP_016884518.1 | |||
| XM_017029030.1 | XP_016884519.1 | |||
| XM_017029031.1 | XP_016884520.1 | |||
| XM_017029032.1 | XP_016884521.1 | |||
| XM_017029033.1 | XP_016884522.1 | |||
| XM_017029034.1 | 16 | 3335 | XP_016884523.1 | |
| AB002369.1 | ||||
| AF233436.2 | AAF40203.2 | |||
| AF233437.1 | AAF40204.1 | |||
| AF233438.1 | AAF40205.1 | |||
| AK289581.1 | ||||
| BC142713.1 | ||||
| BC148216.1 | ||||
| BC151217.1 | ||||
| BC152455.1 | ||||
| CR456525.1 | CAG30411.1 |
More Information
Set Membership: |
|

Genomic Map