Assay Details
Target Gene Details
Entrez Gene ID: | 2147 |
Gene Name: | coagulation factor II, thrombin |
Gene Aliases: |
PT, RPRGL2, THPH1 |
Location: |
Chr.11:46719166-46739508 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2 | NM_000506.4 | 7 | 819 | NP_000497.1 |
| NM_001311257.1 | 7 | 771 | NP_001298186.1 | |
| AJ972449.1 | 7 | 780 | CAJ01369.1 | |
| AK222775.1 | 7 | 754 | BAD96495.1 | |
| AK222777.1 | 7 | 754 | BAD96497.1 | |
| AK293326.1 | 7 | 693 | ||
| AK303747.1 | 7 | 733 | ||
| AK312965.1 | 7 | 784 | ||
| BC051332.1 | 7 | 760 | AAH51332.1 | |
| V00595.1 | 7 | 730 | CAA23842.1 |
More Information
Set Membership: |
|

Genomic Map