Assay Details
Target Gene Details
Entrez Gene ID: | 8398 |
Gene Name: | phospholipase A2 group VI |
Gene Aliases: |
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta |
Location: |
Chr.22:38111495-38192099 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss |
|
| nsv3623 | Chr.22:38105535 - 38113801 on Build GRCh38 | Insertion |
|
| esv996111 | Chr.22:38108982 - 38112324 on Build GRCh38 | Insertion |
|
| nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss |
|
| nsv508732 | Chr.22:38096355 - 38180191 on Build GRCh38 | Insertion |
|
| nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map