Assay Details
Target Gene Details
Entrez Gene ID: | 3356 |
Gene Name: | 5-hydroxytryptamine receptor 2A |
Gene Aliases: |
5-HT2A, HTR2 |
Location: |
Chr.13:46831542-46897076 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR2A | NM_000621.4 | 4 | 2326 | NP_000612.1 |
| NM_001165947.2 | 3 | 1586 | NP_001159419.1 | |
| AK302787.1 | 3 | 1425 | ||
| BC069356.1 | 3 | 1674 | AAH69356.1 | |
| BC069576.1 | 3 | 1674 | AAH69576.1 | |
| BX647777.1 | 3 | 1370 | ||
| X57830.1 | 3 | 1740 | CAA40963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027 | Chr.13:46802456 - 46835423 on Build GRCh38 | Insertion |
|
| esv2763019 | Chr.13:46774094 - 47549312 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map