Assay Details
Target Gene Details
Entrez Gene ID: | 319 |
Gene Name: | apolipoprotein F |
Gene Aliases: |
Apo-F, LTIP |
Location: |
Chr.12:56360571-56362799 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOF | NM_001638.2 | NP_001629.1 | ||
| AK314892.1 | ||||
| BC026257.1 | AAH26257.1 | |||
| CR936663.1 | ||||
| L27050.1 | AAA65642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map