Assay Details
Target Gene Details
Entrez Gene ID: | 2572 |
Gene Name: | glutamate decarboxylase 2 |
Gene Aliases: |
GAD65 |
Location: |
Chr.10:26216307-26304562 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAD2 | NM_000818.2 | NP_000809.1 | ||
| NM_001134366.1 | NP_001127838.1 | |||
| AK127375.1 | BAC86947.1 | |||
| BC029517.1 | AAH29517.1 | |||
| BC126327.1 | ||||
| BC126329.1 | ||||
| M74826.1 | AAA58491.1 | |||
| M81882.1 | AAA62367.1 | |||
| X69936.1 |
More Information
Set Membership: |
|

Genomic Map