Assay Details
Target Gene Details
Entrez Gene ID: | 9245 |
Gene Name: | glucosaminyl (N-acetyl) transferase 3, mucin type |
Gene Aliases: |
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM |
Location: |
Chr.15:59611783-59620011 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT3 | NM_004751.2 | 3 | 1244 | NP_004742.1 |
| AF038650.1 | 1 | 795 | AAD21525.1 | |
| AF102542.1 | 2 | 1148 | AAD10824.1 | |
| AK312852.1 | 2 | 1027 | ||
| BC017032.1 | 3 | 1103 | AAH17032.1 | |
| EF152283.1 | 3 | 1385 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| esv3636644 | Chr.15:59550375 - 59641686 on Build GRCh38 | Loss |
|
| esv2668729 | Chr.15:59550283 - 59641656 on Build GRCh38 | Deletion |
|
| nsv569605 | Chr.15:59563027 - 59664354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map