Assay Details
Target Gene Details
Entrez Gene ID: | 27429 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
OMI, PARK13, PRSS25 |
Location: |
Chr.2:74529405-74533556 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA2 | NM_001321727.1 | 2 | 1159 | NP_001308656.1 |
| NM_001321728.1 | 2 | 1159 | NP_001308657.1 | |
| NM_013247.4 | 2 | 1159 | NP_037379.1 | |
| NM_145074.2 | 2 | 1159 | NP_659540.1 | |
| NR_135769.1 | 2 | 1159 | ||
| NR_135770.1 | 3 | 627 | ||
| NR_135771.1 | 3 | 611 | ||
| NR_135772.1 | 3 | 631 | ||
| AF020760.2 | 2 | 574 | AAB94569.2 | |
| AF141305.1 | 2 | 1159 | AAF66596.1 | |
| AF141306.1 | 2 | 518 | AAF66597.1 | |
| AF141307.1 | 2 | 395 | AAF66598.1 | |
| AF184911.1 | 2 | 574 | AAG13126.1 | |
| AK225932.1 | 2 | 592 | ||
| AK291977.1 | 2 | 578 | ||
| AK310163.1 | 1 | 58 | ||
| BC000096.2 | 2 | 585 | AAH00096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map