Assay Details
Target Gene Details
Entrez Gene ID: | 5476 |
Gene Name: | cathepsin A |
Gene Aliases: |
GLB2, GSL, NGBE, PPCA, PPGB |
Location: |
Chr.20:45890144-45898820 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSA | NM_000308.3 | NP_000299.2 | ||
| NM_001127695.2 | NP_001121167.1 | |||
| NM_001167594.2 | NP_001161066.1 | |||
| NR_133656.1 | ||||
| AB209705.2 | BAD92942.1 | |||
| AK097786.1 | ||||
| AK172808.1 | ||||
| AK304536.1 | ||||
| AK307701.1 | ||||
| AK312898.1 | ||||
| BC000597.2 | AAH00597.1 | |||
| BC093009.1 | AAH93009.1 | |||
| M22960.1 | AAA36476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517296 | Chr.20:45854586 - 45965936 on Build GRCh38 | Loss |
|
| nsv833991 | Chr.20:45885631 - 46068708 on Build GRCh38 | Loss |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map