Assay Details
Target Gene Details
Entrez Gene ID: | 11166 |
Gene Name: | SRY-box 21 |
Gene Aliases: |
SOX25 |
Location: |
Chr.13:94709622-94712543 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX21 | NM_007084.3 | 1 | 1605 | NP_009015.1 |
| BM783159.1 | 1 | 69 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| esv28079 | Chr.13:94710850 - 94713584 on Build GRCh38 | Loss |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
| nsv826750 | Chr.13:94706248 - 94715816 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map