Assay Details
Target Gene Details
Entrez Gene ID: | 10466 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
CDG2I, GOLTC1, GTC90 |
Location: |
Chr.7:107201743-107564514 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG5 | NM_006348.3 | 22 | 4226 | NP_006339.3 |
| NM_181733.2 | 21 | 4163 | NP_859422.2 | |
| AU117533.1 | 1 | 695 |
Target Gene Details
Entrez Gene ID: | 26959 |
Gene Name: | HMG-box transcription factor 1 |
Gene Aliases: |
- |
Location: |
Chr.7:107168961-107202529 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HBP1 | NM_001244262.1 | 11 | 2594 | NP_001231191.1 |
| NM_012257.3 | 11 | 2698 | NP_036389.2 | |
| XM_005250266.2 | 12 | 3184 | XP_005250323.1 | |
| XM_005250267.2 | 11 | 2885 | XP_005250324.1 | |
| XM_017011967.1 | 12 | 2850 | XP_016867456.1 | |
| AF019214.1 | 10 | 2527 | AAB71862.1 | |
| AK025284.1 | 6 | 1819 | ||
| BC017069.1 | 11 | 2698 | AAH17069.1 | |
| BC022329.2 | 10 | 2527 | AAH22329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831087 | Chr.7:107033020 - 107232015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map