Assay Details
Target Gene Details
Entrez Gene ID: | 92154 |
Gene Name: | metastasis suppressor 1 like |
Gene Aliases: |
ABBA-1, ABBA1 |
Location: |
Chr.16:70661204-70686207 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTSS1L | NM_138383.2 | 15 | 2795 | NP_612392.1 |
| XM_005256236.4 | 14 | 2846 | XP_005256293.2 | |
| XM_011523450.2 | 17 | 2756 | XP_011521752.1 | |
| XM_011523453.2 | 16 | 3008 | XP_011521755.2 | |
| XM_011523454.2 | 15 | 2999 | XP_011521756.2 | |
| XM_011523458.2 | 10 | 2233 | XP_011521760.1 | |
| XM_017023879.1 | 16 | 2912 | XP_016879368.1 | |
| XM_017023880.1 | 14 | 2876 | XP_016879369.1 | |
| XM_017023881.1 | 15 | 2855 | XP_016879370.1 | |
| BC002770.1 | AAH02770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
| nsv517391 | Chr.16:70632507 - 70806587 on Build GRCh38 | Loss |
|
| nsv572945 | Chr.16:70632819 - 70806587 on Build GRCh38 | Gain |
|
| nsv471094 | Chr.16:70660097 - 70716067 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map