Assay Details
Target Gene Details
Entrez Gene ID: | 6576 |
Gene Name: | solute carrier family 25 member 1 |
Gene Aliases: |
CTP, D2L2AD, SEA, SLC20A3 |
Location: |
Chr.22:19175575-19178863 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A1 | NM_001256534.1 | NP_001243463.1 | ||
| NM_001287387.1 | NP_001274316.1 | |||
| NM_005984.4 | NP_005975.1 | |||
| NR_046298.2 | ||||
| AK292313.1 | ||||
| AK298204.1 | ||||
| BC004980.1 | AAH04980.1 | |||
| BC008061.2 | AAH08061.1 | |||
| BG473148.1 | ||||
| BI159897.1 | ||||
| BI160240.1 | ||||
| BI560375.1 | ||||
| BP265446.1 | ||||
| DR006840.1 | ||||
| HM037273.1 | ||||
| L75823.1 | AAL40090.1 | |||
| L77567.1 | ||||
| U25147.1 | AAB08515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588213 | Chr.22:18971102 - 19298293 on Build GRCh38 | Gain |
|
| nsv834122 | Chr.22:19098491 - 19280189 on Build GRCh38 | Loss |
|
| esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain |
|
| nsv588216 | Chr.22:19086099 - 19839117 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map