Assay Details
Target Gene Details
Entrez Gene ID: | 158833 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 1 |
Gene Aliases: |
DGA2, DGAT2L3 |
Location: |
Chr.X:70233489-70240661 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AWAT1 | NM_001013579.2 | 1 | 63 | NP_001013597.1 |
| XM_017029295.1 | 1 | 1229 | XP_016884784.1 | |
| AY947638.1 | AAX48018.1 | |||
| W68831.1 | 1 | 63 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522033 | Chr.X:70233184 - 70245422 on Build GRCh38 | Gain |
|
| nsv520416 | Chr.X:70190042 - 70244558 on Build GRCh38 | Loss |
|
| esv3574035 | Chr.X:70227213 - 70246652 on Build GRCh38 | Loss |
|
| nsv527253 | Chr.X:70219663 - 70244558 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map