Assay Details
Target Gene Details
Entrez Gene ID: | 6602 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 |
Gene Aliases: |
BAF60A, CRACD1, Rsc6p |
Location: |
Chr.12:50085200-50100711 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCD1 | NM_003076.4 | NP_003067.3 | ||
| NM_139071.2 | NP_620710.2 | |||
| XM_005269107.3 | XP_005269164.2 | |||
| XM_011538695.1 | XP_011536997.1 | |||
| AF109733.1 | AAD23390.1 | |||
| AK130517.1 | ||||
| AK292700.1 | ||||
| AK293932.1 | ||||
| BC009368.2 | ||||
| CN272379.1 | ||||
| DB010252.1 | ||||
| U66617.1 | AAC50695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558840 | Chr.12:50056193 - 50130181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map