Assay Details
Target Gene Details
Entrez Gene ID: | 55008 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 |
Gene Aliases: |
- |
Location: |
Chr.4:88378565-88443097 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HERC6 | NM_001165136.1 | 1 | 301 | NP_001158608.1 |
| NM_017912.3 | 1 | 301 | NP_060382.3 | |
| XM_005263083.3 | 1 | 462 | XP_005263140.1 | |
| XM_011532053.2 | 1 | 464 | XP_011530355.1 | |
| XM_017008333.1 | XP_016863822.1 | |||
| AF336798.1 | 1 | 301 | AAQ14893.1 | |
| AK092403.1 | 1 | 49 | BAC03879.1 | |
| AK295832.1 | 1 | 211 | ||
| AY653201.1 | 1 | 301 | AAV66895.1 | |
| AY653202.1 | 1 | 301 | AAV66896.1 | |
| AY653203.1 | 1 | 301 | AAV66897.1 | |
| BC042047.1 | 1 | 177 | AAH42047.2 | |
| BX647121.1 | CAI46056.1 | |||
| DA221524.1 | ||||
| DC344051.1 | 2 | 260 |
More Information
Set Membership: |
|

Genomic Map