Assay Details
Target Gene Details
Entrez Gene ID: | 6491 |
Gene Name: | SCL/TAL1 interrupting locus |
Gene Aliases: |
MCPH7, SIL |
Location: |
Chr.1:47250139-47314787 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
More Information
Set Membership: |
|

Genomic Map