Assay Details
Target Gene Details
Entrez Gene ID: | 255877 |
Gene Name: | B-cell CLL/lymphoma 6B |
Gene Aliases: |
BAZF, ZBTB28, ZNF62 |
Location: |
Chr.17:7023050-7029642 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL6B | NM_181844.3 | 3 | 368 | NP_862827.1 |
| AB076580.1 | 3 | 348 | BAC00962.1 | |
| AK122893.1 | 3 | 364 | ||
| AK292878.1 | 3 | 351 | ||
| AK293102.1 | 3 | 367 | ||
| BC059404.1 | 3 | 369 | AAH59404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952117 | Chr.17:7014482 - 7028381 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map