Assay Details
Target Gene Details
Entrez Gene ID: | 120227 |
Gene Name: | cytochrome P450 family 2 subfamily R member 1 |
Gene Aliases: |
- |
Location: |
Chr.11:14877436-14898913 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2R1 | NM_024514.4 | 4 | 1145 | NP_078790.2 |
| XM_005252788.1 | 5 | 1435 | XP_005252845.1 | |
| XM_005252789.2 | 5 | 1688 | XP_005252846.1 | |
| XM_011519895.1 | 5 | 1839 | XP_011518197.1 | |
| XM_011519898.2 | 4 | 7469 | XP_011518200.1 | |
| XM_017017190.1 | 4 | 4030 | XP_016872679.1 | |
| XM_017017191.1 | 4 | 1650 | XP_016872680.1 | |
| XM_017017192.1 | 5 | 1635 | XP_016872681.1 | |
| XM_017017193.1 | 4 | 1597 | XP_016872682.1 | |
| XM_017017194.1 | 4 | 1450 | XP_016872683.1 | |
| XM_017017195.1 | 4 | 3193 | XP_016872684.1 | |
| AK092584.1 | ||||
| AW138676.1 | ||||
| AY323817.1 | 4 | 1145 | AAQ23114.1 | |
| BC104907.1 | 4 | 1190 | AAI04908.1 | |
| BC104909.1 | 4 | 1192 | AAI04910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625429 | Chr.11:14852130 - 14898409 on Build GRCh38 | Gain |
|
| esv3891931 | Chr.11:14839971 - 14950061 on Build GRCh38 | Loss |
|
| esv3578250 | Chr.11:14350967 - 14976647 on Build GRCh38 | Gain |
|
| nsv832071 | Chr.11:14854876 - 15027624 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map